89032007 Sigma

PA317

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, O-R More...
biological source   embryo from mouse
growth mode   Adherent
morphology   Fibroblast
shipped in   dry ice
storage temp.   −196°C

Description

Application

Transfection and virus studies: herpes virus

General description

For complete product information, please see PA317

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C; Grow in HAT for 5 days followed by 4 days in HT once a month and prior to cryopreservation

Cell Line Description

The cell line PA317 was generated by co-transfecting NIH 3T3 TK- cells with retrovirus packaging construct DNA (pPAM3) and the herpes simplex virus thymidine kinase (TK) gene carried as a fragment in pBR322. The construct pPAM3 contains several mutations in addition to deletion of the packaging signal. These features appear to prevent the production of helper virus and the transfer of packaging function. Transfection or infection with retroviral vectors results in the production of retrovirus virions with an amphotropic host range. These are capable of infecting mammalian cells and have been applied to transfer genes into humans. PA317 cells will loose their packaging ability during long-term culture. Therefore, a selection should be included routinely and before cryopreservation of cells. The selection procedure includes growth in HAT-containing medium (0.03mM hypoxanthine, 0.001mM amethopterin, 0.02mM thymidine) for 5 days. To dilute residual amethopterin cells should then be cultured for 4 days in HT containing medium (0.03mM hypoxanthine, 0.02mM thymidine). Cells are stable for at least 1 month after selection.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?