• USA Home
  • 89090402 - AGS Cell Line human

EMAIL THIS PAGE TO A FRIEND
89090402 Sigma

AGS Cell Line human

Purchase

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   stomach from human
description   Human Caucasian gastric adenocarcinoma
growth mode   Adherent
karyotype   Hyperdiploid
morphology   Not specified
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Application

Anti-tumour testing

Cell Line Origin

Human Caucasian gastric adenocarcinoma

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 12
D16S539: 11,13
D5S818: 9,12
D7S820: 10,11
THO1: 6,7
TPOX: 11,12
vWA: 16,17

Culture Medium

Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 0.05% EDTA; 5% CO2; 37°C.

Cell Line Description

Derived from an adenocarcinoma of the stomach of a 54 year-old Caucasian female with no prior anti-cancer treatment. AGS cells are persistently infected with parainfluenza virus type 5 (PIV5).

Price and Availability


Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

recommended

Product #

Description

Add to Cart

G7513 L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?