• USA Home
  • 89468 - Lithium sulfate 0.1M – N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Polyethylene glycol 4000 12% (w/v) – 2-Propanol 2 % (v/v) solution

EMAIL THIS PAGE TO A FRIEND
89468 Sigma

Lithium sulfate 0.1M – N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Polyethylene glycol 4000 12% (w/v) – 2-Propanol 2 % (v/v) solution

Synonym: Crystallization solution 23/Kit-No 73513

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   6.5±0.2 ( in neat)
density   1.04 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?