• USA Home
  • 89754 - Calcium chloride 0.02M – Sodium acetate 0.1M – 2-methyl-2,4-pentanediol 30% solution

EMAIL THIS PAGE TO A FRIEND
89754 Sigma

Calcium chloride 0.02M – Sodium acetate 0.1M – 2-methyl-2,4-pentanediol 30% solution

Synonym: Crystallization solution 01/Kit-No 82009

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   4.6±0.2 (25 °C in neat)
density   1.00 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?