EMAIL THIS PAGE TO A FRIEND
90070501 Sigma

A2B5 clone 105

Reactivity: Anti-Retinal Neuron Glycolipid, chick

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse x Mouse Hybridoma
ImmunoDon   BALB/c mouse spleen
Myeloma   P3X63Ag8
Reactivity   Anti-Retinal Neuron Glycolipid, chick
application(s)   cell culture | mammalian: suitable
isotype   IgM
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

Chicken embryo retina cells

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse P3X63Ag8 x Mouse BALB/c spleen

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

This cytotoxic antibody is directed against a glycolipoid present on chick embryo retinal neuron cell bodies. The antigen is found in chick embryo brain, spinal cord and dorsal root ganglia neurons and in bovine and human brain. The antigen is not present on axons or dendrites.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?