EMAIL THIS PAGE TO A FRIEND
90110605 Sigma

DA6.147

Reactivity: Anti-MHC class II α-chain, human

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse x Mouse Hybridoma
ImmunoDon   BALB/c mouse spleen
Myeloma   NS1
Reactivity   Anti-MHC class II α-chain, human
application(s)   cell culture | mammalian: suitable
isotype   IgG1
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

Daudi Burkitt lymphoma cells

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse NS1 x Mouse BALB/c spleen

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

The antibody is directed against the α chain of human MHC class II antigens. The antibody is poorly reactive with cell surface MHC antigens, but shows excellent reactivity against α chains by Western blotting.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

R0883 RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?