EMAIL THIS PAGE TO A FRIEND
90110606 Sigma

DA6.231

Reactivity: Anti-MHC class II β-chain, human

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse x Mouse Hybridoma
ImmunoDon   BALB/c mouse spleen
Myeloma   NS1
Reactivity   Anti-MHC class II β-chain, human
application(s)   cell culture | mammalian: suitable
isotype   IgG1
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

Daudi Burkitt lymphoma cells

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse NS1 x Mouse BALB/c spleen

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

The antibody is directed against the β chain of human MHC class II antigens. It reacts with cell surface MHC antigens and cross reacts with MHC Class II antigens of other species.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

R0883 RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?