90112714 Sigma

PC-3 Cell Line human

Caucasian prostate adenocarcinoma

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, O-R More...
biological source   prostate from human
growth mode   Adherent
karyotype   2n = 46, triploid, modal no. 62
morphology   Epithelial
shipped in   dry ice
storage temp.   −196°C

Description

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11
D16S539: 11
D5S818: 13
D7S820: 8,11
THO1: 6,7
TPOX: 8,9
vWA: 17

Culture Medium

Coons Modified Ham’s F12 + 2mM Glutamine + 7% Foetal Bovine Serum (FBS).OR Kaign’s modified Ham’s F12 + 45mg/L ascorbic acid + 18 mg/L Inositol + 2mM Glutamine + 7% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Medium change every 5 days. The initial subculture interval after cells are thawed may be longer than 7-9 days.

Cell Line Description

Established from a grade 4 prostatic adenocarcinoma from a 62 year old male Caucasian. The cells grow in agar and produce tumours in nude mice. Exhibit low acid phosphatase and testosterone-5-α reductase activity. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Customers Also Viewed

prostate carcinoma

breast adenocarcinoma

human breast adenocarcinoma

95012614PNT1A human

Normal prostate epithelium imortalized with SV40.

Normal prostate epithelium immortalized with SV40.

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?