• USA Home
  • 90455 - Triethylene glycol monooctyl ether

EMAIL THIS PAGE TO A FRIEND
90455 Sigma

Triethylene glycol monooctyl ether

BioXtra, ≥98.0% (GC)

Synonym: C8E3, Octyl triethylene glycol ether, Octyltriglycol

  • CAS Number 19327-38-9

  • Linear Formula CH3(CH2)7(OCH2CH2)3OH

  • Molecular Weight 262.39

  •  Beilstein Registry Number 1765631

  •  MDL number MFCD00043034

  •  PubChem Substance ID 24889369

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
InChI Key   XIVLVYLYOMHUGB-UHFFFAOYSA-N
product line   BioXtra
assay   ≥98.0% (GC)
refractive index   n20/D 1.445
density   0.948 g/mL at 20 °C(lit.)
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cr: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤50 mg/kg
  Mg: ≤50 mg/kg
  Mn: ≤50 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

Useful for solubilising membrane proteins

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?