• USA Home
  • 90877 - N-[trans-4-(Succinimidyloxycarbonyl)cyclohexylmethyl]-sulforhodamine B-acid amide

EMAIL THIS PAGE TO A FRIEND
90877 Sigma

N-[trans-4-(Succinimidyloxycarbonyl)cyclohexylmethyl]-sulforhodamine B-acid amide

suitable for fluorescence, ≥85.0% (coupling to amines)

  • Empirical Formula (Hill Notation) C39H46N4O10S2

  • Molecular Weight 794.93

  •  PubChem Substance ID 329769960

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Luminescent Compounds/Detection, Rhodamine and Derivatives,
InChI Key   PMEJVCSFSLFHAP-DIVCQZSQSA-N
assay   ≥85.0% (coupling to amines)
solubility   DMF: soluble
fluorescence   λex ≥566 nm; λem ≥587 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?