EMAIL THIS PAGE TO A FRIEND
90917 Sigma

ZnAF-1F

≥90% (HPCE)

Synonym: 5-{2-[Bis(2-pyridylmethyl)amino]ethylamino}-2′,7′-difluorofluorescein

Purchase

Properties

Related Categories Biochemicals and Reagents, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains, Zinc Probes More...
InChI Key   PUDBCJSXTISPOO-UHFFFAOYSA-N
assay   ≥90% (HPCE)
fluorescence   λex 492 nm; λem 517 nm in PBS, Zn2+

Description

Application

suitable for the fluorimetric detection of Zn2+

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?