91031103 Sigma

MPC-11

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, J-N More...
biological source   Blood from mouse
growth mode   Suspension
karyotype   Modal no. 67-70
morphology   Lymphoblast
products   Immunoglobulin G (IgG2b), half molecules, light chain dimers, free light chains
shipped in   dry ice
storage temp.   −196°C

Description

General description

For complete product information, please see MPC-11

Culture Medium

DMEM + 2mM Glutamine + 20% Horse Serum (HS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

Established from the Merwin plasma cell tumour-II carried in BALB/c mice. Resembles the parent tumor in that they both produce fully assembled gamma globulin molecules (IgG2b) as well as half molecules, light chain dimers and free light chains. The MPC-11 produces 5-6 picograms of IgG2b per cell per minute.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Customers Also Viewed

2-<WBR>Methacryloyloxyethyl phosphorylcholine

contains ≤100 ppm 4-methylphenol as inhibitor, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?