| Related Categories | Cell Culture, ECACC Cell Lines, S-Z More... |
| biological source | Ovary from human |
| description | Human Caucasian ovary adenocarcinoma |
| growth mode | Adherent |
| karyotype | Hypodiploid to hypotetraploid |
| morphology | Epithelial |
| shipped in | dry ice |
| storage temp. | −196°C |
STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 8,11
D16S539: 12
D5S818: 11
D7S820: 13,14
THO1: 9,9.3
TPOX: 8,11
vWA: 17,18
McCoy′s 5a + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-6x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Derived from the ascitic fluid from a 64 year old caucasian female with an ovarian tumour. Forms moderately well-differentiated adenocarcinoma consistent with ovarian primary cells.
Customers Also Viewed
ovarian carcinoma
Modified, with
breast adenocarcinoma
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?