EMAIL THIS PAGE TO A FRIEND
91335 Sigma

Pullulan

Purchase

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Pullulan (PULL), a glucan polymer composed of α(1,6) linked maltotriose (three glucose) units, is used for the development of copolymers, permeability barrier coatings, nanofibers, nanospheres, nanogels, and biofilms with potential uses that range from protective food wrapping materials to biocompatible tissue engineering and drug delivery materials and structures.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Prebiotics

Prebiotic food components typically consist of polysaccharides and oligosaccharides. They can be found in milk (galactooligosaccharides or GOS) and plants with dietary fibers, and they are often prod...
BioFiles Edition 6.2
Keywords: Fermentation, Prebiotics

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?