• USA Home
  • 91517 - Fluorescent red NIR 782

EMAIL THIS PAGE TO A FRIEND
91517 Sigma

Fluorescent red NIR 782

suitable for fluorescence

  • Empirical Formula (Hill Notation) C38H48N2O6S

  • Molecular Weight 660.86

  •  MDL number MFCD03792251

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Fluorescent Red and Orange Labels, Luminescent Compounds/Detection,
solubility   DMSO: soluble
fluorescence   λex 635 nm; λem ~800 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
storage temp.   2-8°C

Description

Analysis Note

further Exc.: 782 nm (results in a lower emission)

General description

Fluorescenct Red NIR 782 is a new fluorescent dye expecially well suited for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required. Fluorescence NIR 782 shows strong fluorescence.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?