EMAIL THIS PAGE TO A FRIEND
91950 Sigma

Trihydroxyethylrutin

≥80% (HPLC)

Synonym: 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Troxerutin

  • CAS Number 7085-55-4

  • Empirical Formula (Hill Notation) C33H42O19

  • Molecular Weight 742.68

  •  Beilstein Registry Number 4778232

  •  EC Number 230-389-4

  •  MDL number MFCD00893813

  •  PubChem Substance ID 57653462

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Natural Dyes, Stains and Dyes
InChI Key   IYVFNTXFRYQLRP-VVSTWUKXSA-N
assay   ≥80% (HPLC)
optical activity   [α]20/D −48±1°, c = 1% in pyridine

Description

Biochem/physiol Actions

Shows widespread protective effects against oxidative DNA damage induced by D-galactose as well as other types of oxidative stress.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LK8331500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?