92020424 Sigma

MDA-MB-231 Cell Line human

human breast adenocarcinoma

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, J-N More...
biological source   Breast from human
growth mode   Adherent
karyotype   Modal no.′s 62 and 64, near triploid
morphology   Epithelial
shipped in   dry ice
storage temp.   −196°C

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Application

MDA-MB-231 was isolated from pleural effusions of a Caucasian breast cancer patient.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12,13
D13S317: 13
D16S539: 12
D5S818: 12
D7S820: 8,9
THO1: 7,9.3
TPOX: 8,9
vWA: 15,18

Culture Medium

L15+ 2mM Glutamine + 15% Foetal Bovine SerumFBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e. seeding at 3-6x10,000 cells/cm2 using 0.25% Trypsin or Trypsin/EDTA; 37°C. CO2 is not required.

Cell Line Description

Isolated from pleural effusions of a breast cancer patient.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Customers Also Viewed

With L-glutamine, powder, suitable for cell culture

Without L-glutamine, liquid, sterile-filtered, suitable for cell culture

With L-glutamine, liquid, sterile-filtered, suitable for cell culture

breast adenocarcinoma

92020422MDA-MB-157

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?