• USA Home
  • 92090213 - T98G Cell Line human

EMAIL THIS PAGE TO A FRIEND
92090213 Sigma

T98G Cell Line human

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   brain from human
description   Human Caucasian glioblastoma
growth mode   Adherent
karyotype   Hyperpentaploid, modal no. 128-132
morphology   Not specified
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Application

Studies on the mechanism for cessation of proliferation in quiescent cells and cells synchronised in G1 phase.

T98G has been used to study the effects of the lysosomal destabilizing drug Siramesine on glioblastoma. It has also been used to study the efficiency of empty spiracles homeobox 2 (EMX2) in suppressing glioblastoma.

Cell Line Origin

Human Caucasian glioblastoma

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 13
D16S539: 13
D5S818: 10,12
D7S820: 9,10
THO1: 7,9.3
TPOX: 8
vWA: 17,20

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1% Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Cells may become anchorage-independent if deprived of serum.

Cell Line Description

Derived from glioblastoma multiform tumour from a 61-year-old Caucasian male. Indefinite lifespan and anchorage independent but can enter viable G1 arrested state when deprived of serum.

Price and Availability


Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
  What kind of solution is the A5955 - Antibiotic Antimycotic Solution (100×), Stabilized product suspended in?
A5955 - Antibiotic Antimycotic Solution (100×), Stabilized is supsended in a sodium citrate buffer solution. Source: Israel production group
Show more questions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

M7145 MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
M2279 Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
G7513 L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
S8636 Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?