• USA Home
  • 92333 - N-(4-Methylumbelliferyl)maleimide

EMAIL THIS PAGE TO A FRIEND
92333 Sigma

N-(4-Methylumbelliferyl)maleimide

Synonym: 7-Maleimido-4-methylcoumarin

  • CAS Number 211565-47-8

  • Empirical Formula (Hill Notation) C14H9NO4

  • Molecular Weight 255.23

  •  MDL number MFCD11114065

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Luminescent Compounds/Detection, Other Fluorescent Labels,
InChI Key   SKWQLGLJKVPHQT-UHFFFAOYSA-N
fluorescence   λex 330 nm; λem 393 nm in 0.1 M phosphate pH 7.0, 2-mercaptoethanol
suitability   corresponds for coupling to thiols
storage temp.   2-8°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?