| Related Categories | Cell Culture, ECACC Cell Lines, O-R More... |
| biological source | embryo from rat |
| growth mode | Adherent |
| karyotype | Near diploid |
| morphology | Fibroblast |
| shipped in | dry ice |
| storage temp. | −196°C |
For complete product information, please see R9
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 5% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
The R74 line was derived by co-cultivation of 18 day-old Lewis rat embryonic fibroblasts with cells of a chicken positive for Rous sarcoma virus-33 (RSV-33). The R9 cell population is clearly distinguishable from RT1 and R74 cell lines because it contains flat polygonal and triangular refractile cells with a tendency to round up. It is also tumourigenic in chickens and rats.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?