• USA Home
  • 93296 - TRIS acetate – EDTA buffer solution

EMAIL THIS PAGE TO A FRIEND
93296 Sigma

TRIS acetate – EDTA buffer solution

for molecular biology, concentrate, 10× concentrate

Synonym: TAE buffer solution

Purchase

Properties

grade   for molecular biology
quality   10× concentrate
impurities   DNases, none detected
  RNases, none detected
  phosphatases, none detected
  proteases, none detected
density   1.02 g/mL at 20 °C
cation traces   Al: ≤1 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤1 mg/kg
  Bi: ≤1 mg/kg
  Ca: ≤5 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤20 mg/kg
  Li: ≤1 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Mo: ≤1 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Sr: ≤1 mg/kg
  Zn: ≤1 mg/kg
suitability   in accordance for electrophoresis test

Description

Physical form

0.4 M TRIS acetate, pH 8.3±0.2 (25°C); 0.01 M EDTA.Na2;

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?