• USA Home
  • 93379 - Trizma® Buffer substance

EMAIL THIS PAGE TO A FRIEND
93379 Sigma

Trizma® Buffer substance

pH 8.1

Synonym: Tris Buffer substance, Tris(hydroxymethyl)aminomethane Buffer substance

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffers A to Z, Good’s Buffers
InChI Key   QKNYBSVHEMOAJP-UHFFFAOYSA-N
mol wt   average mol wt 139.8
pH   7.8±0.1 (37 °C, 0.05 M)
  8.1±0.1 (25 °C, 0.05 M)
  8.7±0.1 (5 °C, 0.05 M)
solubility   H2O: soluble0.67 g/mL, clear, colorless

Description

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?