EMAIL THIS PAGE TO A FRIEND
93418 Sigma

Triton X-100

BioXtra

Synonym: 4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Purchase

Properties

Related Categories Antigen-Vaccine Preparation, Biochemicals and Reagents, Cell Culture, Detergents, Detergents N to Z,
description   non-ionic
InChI Key   HNLXNOZHXNSSPN-UHFFFAOYSA-N
product line   BioXtra
mol wt   micellar avg mol wt 80,000
  average mol wt 625
aggregation number   100-155
impurities   ≤0.001% peroxides (as H2O2)
  ≤0.5% water
ign. residue   ≤0.2%
CMC   0.2-0.9 mM(20-25°C)
transition temp   cloud point 65 °C
  pour point ~7 °C
solubility   H2O: soluble0.005 M at 20 °C, clear, colorless
density   1.065 g/mL at 20 °C
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤500 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤500 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
HLB   13.5

Description

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Other Notes

Non-ionic surfactant, for membrane solubilization. Can be removed from aqueous solution by Amberlite® XAD-2; Partial purification of the insulin receptor

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
MD0907700
Flash Point(F) 
483.8 °F
Flash Point(C) 
251 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?