• USA Home
  • 93611 - Trypsin, DPCC treated, from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
93611 Sigma

Trypsin, DPCC treated, from bovine pancreas

lyophilized, 6000-12000 U/mg

Synonym: DPCC-Trypsin

Purchase

Properties

Related Categories 3.4.x.x Peptidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Enzyme Class Index,
quality   lyophilized
mol wt   Mr ~23000
foreign activity   chymotrypsin ≤0.001%
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Sales restrictions may apply

Treatment with diphenyl carbamyl chloride (DPCC) reduces chymotrypsin usually present in trypsin; Thia-analogues of amino acids, synthesis of peptide derivatives containing 3-thia-analogues of amino acids

Unit Definition

1 U corresponds to the amount of enzyme which increases the absorbance at 253 nm by 0.001 per minute at pH 7.6 and 25°C (N-benzoyl-L-arginine ethyl ester, Cat. No. 12880, as substrate)

Application

Tyrpsin is a serine protease used to hydrolyze proteins. Trypsins are used for the re-suspension of cells during cell culture and in proteomics research for the digestion of various proteins . Product 93611 is from bovine pancreas and is DPCC treated. Treatment with diphenyl carbamyl chloride (DPCC) reduces chymotrypsin usually present in trypsin, thia-analogues of amino acids and the synthesis of peptide derivatives containing 3-thia-analogues of amino acids.

Biochem/physiol Actions

Tyrpsin cleaves peptide chains, amides and esters mainly at the carboxyl side of the amino acids lysine or arginine. Tyrpsin plays an important role in digestion and protein absorption. Tyrpsin originates in the pancreas in the form of the zymogen, trypsinogen.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YN5075000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?