• USA Home
  • 93619 - Trypsin inhibitor from Glycine max (soybean)

EMAIL THIS PAGE TO A FRIEND
93619 Sigma

Trypsin inhibitor from Glycine max (soybean)

lyophilized, ~12000 U/mg

Synonym: SBTI

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Proteins More...
quality   lyophilized
mol wt   Mr ~20000
shipped in   wet ice
storage temp.   −20°C

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

Prepared by the method of M. Kunitz

Preparation Note

The trypsin inhibitor is soluble in water and phosphate buffers at 10 mg/mL. It is soluble in balanced salt solutions at 1 mg/mL and in serum-free media. Concentrated solutions greater than 10 mg/mL may be hazy and have a yellow to amber color. After trypsinizing cells, resuspend in 1 mL trypsin inhibitor solution at 1 mg/mL for every mL of trypsin solution used for dissociation. The cell suspension should then be centrifuged at 1000 rpm, forming a cell pellet.

Solutions can retain activity when stored short-term at 2-8° C. Solutions are stable in frozen aliquots at -20°C.

Unit Definition

1 U corresponds to the amount of inhibitor which reduces the trypsin activity by 1 BAEE-U. (1 BAEE-U is the amount of enzyme which increases the absorbance at 253 nm by 0.001 per minute at pH 7.6 and 25 °C; BAEE, Cat. No. 12880, as substrate).

One trypsin unit = A253 of 0.001 per minute with N-alpha-benzoyl-L-arginine ethyl ester (BAEE) as substrate at pH 7.6 at 25 °C.

Biochem/physiol Actions

This inhibitor acts against trypsin, and chymotrypsin and plasmin to a lesser extent. It will also inhibit proteases with mechanisms similar to trypsin, plasma kallikrein and coagulation Factor X. The trypsin inhibitor will not act against metalloproteases, tissue-baseed kallikrein, acid proteases, or thio proteases. This inhibitor acts by forming a 1:1 stoichiometric complex with the protease active site, and then cleaving a single arginine-isoleucine bond on the inhibitor. The inhibition is both reversible and pH dependent.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
What is the molecular weight and amino acid sequence of trypsin inhibitor from Glycine max (soybean)?    
Soybean trypsin inhibitor is a mixture of the three types. We have not characterized our trypsin inhibitor products in terms of how much of each of the three types may be present. All three have a molecular weight of between 20,000 and 20,200 daltons, so one might use 20,100 daltons as an average. The three types vary in the amino acids at a few positions. The detailed summary of the three types (or variants) was published in the following linked reference:      J. Biochem. (Japan), 98, 435-448 (1985). Note that the authors use Tia, Tib and Tic to indicate the three variants.
Show more questions
Protocols & Articles

Articles

HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)
Keywords: Chromatography, High performance liquid chromatography, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?