• USA Home
  • 94285 - Urease from Canavalia ensiformis (Jack bean)

EMAIL THIS PAGE TO A FRIEND
94285 Sigma

Urease from Canavalia ensiformis (Jack bean)

lyophilized, powder, ~100 U/mg

Purchase

Description

Other Notes

Used for the enzymatic determination of urea; Urea microsensor.

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol urea (Cat. No. 51459) per minute at pH 8.0 and 25 °C.

Application

Urease from Canavalia ensiformis may be used for urea determination of various samples, such as legumes . It may be useful for the detection of pathogens as well as heavy-metal ions. Product 94285 is provided as a lyophilized powder (~100 U/mg).

Biochem/physiol Actions

Urease catalyzes the hydrolysis of urea into carbon dioxide and ammonia. Urease is involved in nitrogen metabolism and urea degradation. Urease from Canavalia ensiformis binds 2 nickel ions per subunit .

General description

Subunit molecular weight: ~90,770
Composed of six subunits with total molecular weight: ~544,620

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?