• USA Home
  • 94836 - [Arg8]Vasopressin acetate

EMAIL THIS PAGE TO A FRIEND
94836 Sigma

[Arg8]Vasopressin acetate

≥96.0% (HPLC)

Synonym: AVP, Argipressin acetate, [8-Arginine]Vasopressin acetate

  • CAS Number 129979-57-3

  • Empirical Formula (Hill Notation) C46H65N15O12S2

  • Molecular Weight 1084.23

  •  Beilstein Registry Number 8609690

  •  MDL number MFCD00133939

  •  PubChem Substance ID 57653644

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones, Pituitary Hormones,
InChI Key   ZHFGXNMADRRGQP-HPILINOVSA-N
assay   ≥96.0% (HPLC)
storage temp.   −20°C
Gene Information   human ... AVP(551)

Description

Amino Acid Sequence

Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2 [Disulfide bridge: 1-6]

Other Notes

Thyrotropin-releasing hormone

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?