• USA Home
  • 95576 - BioStab enzyme stabilizer

EMAIL THIS PAGE TO A FRIEND
95576 Sigma

BioStab enzyme stabilizer

Synonym: Enzyme stabilizer BioStab, Stabilizer for Enzymes

Purchase

Properties

Related Categories Biochemicals and Reagents, Biostabilization Solutions, Enzyme Stabilizers, Preformulated Enzyme Stabilizers, Proteins and Derivatives,
impurities   DNases, none detected
  RNases, none detected
  proteases, none detected
storage temp.   2-8°C

Description

Application

A thermostable, chemical inert substance has been isolated from extremophilic microorganisms and purified. This substance stabilizes biological molecules (enzymes). Within the formulation of BioStab enzyme stabilizer it acts similar to its natural function, protecting proteins against denaturation caused by thermal stress, proteolysis, changes in pH or salt concentration.

General description

BioStab enzyme stabilizer is a 4x concentrate in sterile, double-distilled water.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?