Synonym: FITC–DEAE–Dextran 20, Fluorescein isothiocyanate–Diethylaminoethyl–Dextran
| Related Categories | Biochemicals and Reagents, Carbohydrates, Dextrans, Fluoroscein Isothiocyanate (FITC)-DEAE-Dextran Conjugates |
| mol wt | average mol wt 20,000 |
| composition | nitrogen, 2-5% |
| extent of labeling | 0.001-0.008 mol/mol FITC glucose |
| fluorescence | λex 493 nm; λem 518 nm±5 nm in 0.1 M Tris pH 8.0 |
Customers Also Viewed
powder
BioReagent, for molecular biology
average mol wt 70,000
(FITC:Glucose = 1:100)
average mol wt 3,000-5,000
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?