Synonym: 4′-Octyl-4-biphenylcarboxylic acid, 4′-n-octylbiphenyl-4-carboxylic acid, 8BCA
CAS Number 59662-49-6
Empirical Formula (Hill Notation) C21H26O2
Molecular Weight 310.43
MDL number MFCD00157040
PubChem Substance ID 24724405
| Related Categories | A-AM, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Gene Regulation and Expression, |
| assay | ≥98% (HPLC) |
| form | solid |
| solubility | DMSO: >7 mg/mL |
| storage temp. | 2-8°C |
| Gene Information | human ... RARB(5915) |
AC-55649 is a subtype selective RAR (RARβ2) agonist. AC-55649 is a potent, orally available isoform selective Retinoic Acid Receptor β2. AC-55649 has high selectivity, 99% for RARβ2 and 31% for RARβ1. When compared to the gold standard AM-580 (Cat. No A8843), both isoform receptors were 100%. AC-55649 has a potent EC50 of 6.9 μM compared to an EC50 of 7.7 μM for AM-580.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?