MDL number MFCD00130292
| Related Categories | Alphabetical, B, Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Indicators and Probes, |
| mol wt | mol wt ~10,000 |
| extent of labeling | ~1 mol BCECF per mol |
| matrix | dextran |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?