CLS431352 Sigma

Corning® fiberglass prefilters

membrane diam. 90 mm, for 1000mL filter funnels

DOWNLOAD MSDS (PDF)

Synonym: fiber glass filters

Properties

Related Categories Corning, Corning Fiberglass Prefilters, Corning Filters, Corning Plastic Filters, Filtration Supplies,
sterility   non-sterile
packaging   case of 96
  pack of 12
mfr. no.   Corning, 431352
membrane diam.   90 mm

Description

General description

Corning fiberglass prefilters for use with 1000mL filter funnels.

Note: 8 per bag; 12 bags per case

Legal Information

Corning is a registered trademark of Corning, Inc.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?