D5820 Sigma

Anti-DUT (119-130) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

DOWNLOAD MSDS (PDF)

Synonym: Anti-Deoxyuridine 5′-triphosphate nucleotidohydrolase, isoform DUT-N mitochondrial precursor, Anti-dUTPase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to DNA Repair Proteins,
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~18 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... DUT(1854)

Description

Immunogen

synthetic peptide corresponding to amino acids 119-130 of human DUT

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability

Anti-Rabbit 2° Antibody Best Sellers


Prod. No.ConjugateSecondary Antibody Description
A0545HRPAnti-Rabbit IgG (whole molecule)-Peroxidase
A3687APAnti-Rabbit IgG (whole molecule)-Alkaline Phosphatase
F0382FITCAnti-Rabbit IgG (whole molecule)-FITC
B8895BiotinAnti-Rabbit IgG (whole molecule)-Biotin





Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?