Synonym:
2-Phospho-
CAS Number 9014-08-8
EC Number 232-754-3
MDL number MFCD00131019
| Related Categories | 4.2.x.x C-O bonds, 4.x.x.x Lyases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates, |
| form | ammonium sulfate suspension |
| foreign activity | lactic dehydrogenase ≤0.1% |
| pyruvate kinase ≤0.1% | |
| storage temp. | 2-8°C |
Enolase is a metalloenzyme that catalyzes the interconversion of 2-phosphoglycerate to phosphoenolpyruvate. Enolase is essential for both glycolysis and gluconeogenesis.
One unit will convert 1.0 μmole of 2-phosphoglycerate to phospho(enol)pyruvate per min at pH 7.4 at 25 °C.
Crystalline suspension in 2.8 M (NH4)2SO4 containing 0.05 M imidazole and 1 mM MgSO4, pH 7.5
Enolase from rabbit muscle has been used to study its rate-limiting steps and the site of Li+ inhibition.
Customers Also Viewed
lyophilized powder, ≥50 units/mg protein
lyophilized powder, ≥75 units/mg protein
≥90% (SDS-PAGE), buffered aqueous solution
Type II, ammonium sulfate suspension, 350-600 units/mg protein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?