F4550 Sigma
AlumaSeal 96 ® foil
DOWNLOAD MSDS (PDF)
Description
General description
AlumaSeal is ideal for the harshest environments and conditions, cold storage, incubation and transportation. Proprietary adhesive is chemically resistant to DMSO and similar harsh chemicals but is inert and will not leach. Ideal for sealing plates that are light sensitive and it is pierceable to allow for direct sample recovery without peeling. An excellent choice for robotics. Note: Not recommended for PCR applications.
Legal Information
AlumaSeal is a registered trademark of Excel Scientific, Inc.
Price and Availability
Customers Also Viewed
AlumaSeal<SUP>®</SUP> 96 film
size 38 μm, thick aluminum foil sealing film for use with 96 well plates, non-sterile
AlumaSeal<SUP>®</SUP> CS Films for cold storage
size 50 μm, formulated aluminum foil sealing films, non-sterile
AlumaSeal<SUP>®</SUP> CS Films for cold storage
size 50 μm, formulated aluminum foil sealing films, sterile
AlumaSeal<SUP>®</SUP> II film
non-sterile
AlumaSeal<SUP>®</SUP> II film
for PCR and cold storage, sterile
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?