Synonym: Anti-CASP8
MDL number MFCD01633094
Popular Documents: Datasheet (PDF)
| Related Categories | A to L, Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, |
| species reactivity | human |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | buffered aqueous solution |
| shipped in | wet ice |
| storage temp. | −20°C |
| Gene Information | human ... CASP8(841) |
| mfr. no. | Genway, 15-288-21154 |
| NCBI accession no. |
NP_203519.1 |
| Swiss-Prot accession no. |
Q14790 |
Immunogen Sequence: GI # 15718706
, sequence 266-436
Recombinant caspase 8 isoform B
Solution in phosphate buffered saline containing 0.02% sodium azide.
| Prod. No. | Conjugate | Secondary Antibody Description |
| A9046 | HRP | Anti-Chicken IgY (IgG) (whole molecule)-Peroxidase |
| F8888 | FITC | Anti-Chicken IgY (IgG) (whole molecule)-FITC |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?