Synonym: IL-2 sRα
MDL number MFCD01867100
Popular Documents: Datasheet (PDF)
| Related Categories | Cancer Research, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Cytokines & Growth Factors, |
| recombinant | expressed in baculovirus infected Sf21 cells |
| assay | >97% (SDS-PAGE) |
| form | lyophilized powder |
| impurities | endotoxin, tested |
| storage temp. | −20°C |
| Gene Information | human ... IL2RB(3560) |
The bioactivity is measured by its ability to inhibit the IL-15-dependent proliferation of a human megakaryocytic leukemic cell line, M07e.
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.25 mg bovine serum albumin.
Recombinant human IL-2 soluble receptor β is a member of the cytokine receptor superfamily. It binds IL-2 with low affinity but will bind IL-15 sufficiently for neutralization activities.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?