Synonym:
Anti-
MDL number MFCD02263804
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Neurobiology, Antibodies to Neural Proteins, Antibodies to Synaptic Proteins, |
| species reactivity | mouse, human, bovine, rat, chicken |
| application(s) | immunohistochemistry: suitable |
| immunoprecipitation: suitable | |
| indirect immunofluorescence: suitable | |
| western blot: 1:2,000 using PC12, MDBK, 3T3, or normal human fibroblast cells | |
| clone | NSF-1, monoclonal |
| concentration | ~1 mg/mL |
| antibody form | purified immunoglobulin |
| form | buffered aqueous glycerol solution |
| isotype | IgG |
| mol wt | mol wt ~76 kDa |
| shipped in | wet ice |
| storage temp. | −20°C |
| Gene Information |
human ... NSF(4905) mouse ... Nsf(18195) rat ... Nsf(60355) |
recombinant human NSF.
NSF acting in concert with SNAPs (soluble NSF attachmentproteins) and SNARES (SNAP receptors), is requiredfor the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to- endosome fusion and
Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?