P2739 Sigma

Poly(A)+ RNA human

from human normal adult (Uterus)

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Biology, Expression/Translation, Gene Expression & Analysis, General Stem Cell Biology, Molecular Biology,
biological source   from human normal adult (Uterus)
shipped in   dry ice
storage temp.   −70°C
  −70°C

Description

Other Notes

Human mRNA is isolated from total RNA by affinity chromatography using oligo(dT) columns. Following isolation, the integrity of the mRNA is verified on a denaturing agarose gel. All mRNAs show very high efficiencies in reverse transcription.

Packaging

Each vial contains 5 μg of poly(A)+ RNA in DEPC treated water

Legal Information

Manufactured by BioChain, Inc.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?