PQ0221 Sigma

ProteoQwest Chemiluminescent Western Blotting Kit, CPS Substrate

(for Rabbit Antibodies)

DOWNLOAD MSDS (PDF)

Description

Features and Benefits

Benefits include
• Short procedure time
• Reproducible results
• Minimal background

General description

ProteoQwest Chemiluminescent Western Blotting Kit, CPS Substrate, is designed for high sensitivity, low background chemiluminescent detection of as little as 0.06 ng of protein. All of the components of this ProteoQwest kit have been extensively tested and optimized. This kit is designed for 12 mini-gel sized (10 × 10 cm) blots. It is possible to use this kit for as many as 24 blots if half the suggested amount of reagents is used.

Other Notes

Store the reagents as supplied in the kit at 2-8°C, except for the Chemichrome Ultimate, which should be stored in the freezer at -20°C. The unreconstituted packets and reagents are stable for 1 year. Once reconstituted, the Tris-Buffered Saline with TWEEN 20 is stable for 2 weeks at 2-8°C if filtered into a sterile container using a 0.2 μm sterile filter. Once reconstituted, the Tris-Buffered Saline with 3% Milk is stable for 1 day at 2-8°C.

Legal Information

ProteoQwest is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Kit component only

Description

Product #

Add to Cart

Tris Buffered Saline with TWEEN® 20, BioReagent 12 packets    
Tris Buffered Saline, pH 8.0, with 3% nonfat Milk, powder 12 packets    
Anti-Rabbit IgG (whole molecule)-Peroxidase, ProteoQwest antibody produced in goat, affinity isolated antibody, buffered aqueous solution 150 μL    
Chemiluminescent Peroxidase Substrate-1, sufficient for 60 mL substrate 60 mL    

Standard component

Description

Product #

Add to Cart

ColorBurst Electrophoresis Marker, mol wt 8,000-220,000 Da 1VL C1992

Customers Also Viewed

Chemiluminescent Peroxidase Substrate-1

sufficient for 60 mL substrate

ColorBurst<SUP>™</SUP> Electrophoresis Marker

mol wt 8,000-220,000 Da

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?