Popular Documents: Bulletin (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibody Kits, Immunohistology Kits, MO-MR, |
| shipped in | wet ice |
| storage temp. | 2-8°C |
Sigma Rapid Staining Kits are developed for use with high affinity antibodies. The methodology used is based on the Avidin/Biotin detection system, using the uniquely modified avidin reagent, ExtrAvidin®-Peroxidase. The kits are designed as research tools for the identification of antigens in formalin-fixed, paraffin-embedded tissue sections. Labeling is complete in 20 minutes or less. High affinity antibodies must be used with this methodology. Each Rapid Staining Kit contains reagents for ~50 slides and consists of ready-to-use biotinylated secondary antibody, ExtrAvidin®-Peroxidase reagent, substrate buffer, hydrogen peroxide, the chromogen 3-amino-9-ethylcarbazole (AEC) and easy-to-follow instructions.
|
Description |
Product # |
Add to Cart |
|---|---|---|
| instructions |
|
Description |
Product # |
Add to Cart |
|---|---|---|
| 3-amino-9-ethylcarbazole | ||
| biotinylated second antibody | ||
| ExtrAvidin-Peroxidase | ||
| hydrogen peroxide | ||
| substrate buffer |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?