Synonym: 2,4-Hexadienoic acid potassium salt
CAS Number 24634-61-5
Empirical Formula (Hill Notation) C6H7KO2
Molecular Weight 150.22
EC Number 246-376-1
MDL number MFCD00016546
PubChem Substance ID 24899490
| Related Categories | Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antibiotics by Application, Antifungal, |
| assay | ≥99% |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?