Synonym: Anti-Polymerase (DNA directed), delta 2, regulatory subunit 50kDa
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to DNA Repair Proteins, |
| species reactivity | bovine, human, mouse, rat |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
LREVSEEHNLLPQPPRSKYIHPDDE |
| storage temp. | −20°C |
| Gene Information | human ... POLD2(5425) |
Peptide region of the protein sequence according to NP_006221.
Lyophilized from PBS buffer with 2% sucrose
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?