Synonym: Anti-M40, Anti-MGC117247, Anti-MGC16435, Anti-OK/SW-cl.56, Anti-Tubulin, β
MDL number MFCD00145892
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytoskeleton, Antibodies to Microtubule and Associated Proteins, |
| species reactivity | pig, bovine, zebrafish, canine, human, mouse, rat, chicken |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
YHGDSDLQLDRISVYYNEATGGKYV |
| storage temp. | −20°C |
| Gene Information | human ... TUBB(203068) |
Peptide region of the protein sequence according to NP_821133.
Lyophilized from PBS buffer with 2% sucrose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?