Synonym: Anti-Histone H4 (mono methyl K20)
EC Number 231-791-2
| Related Categories | Alphabetical Index, Antibodies, ChIP Validated Antibodies, Epigenetics, H1-HE, |
| species reactivity | human |
| application(s) | ChIP: suitable |
| western blot: suitable | |
| antibody form | culture supernatant |
| form | buffered aqueous solution |
| mol wt | antigen mol wt ~12 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... H4F3(3023) |
| NCBI accession no. |
X60653 |
|
X60653 |
Supplied as concentrated culture supernatant containing 0.05% sodium azide.
Imprint is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?