SHC317V Sigma

MISSION® 1X LacO Inducible Luciferase shRNA Control Transduction Particles

shRNA sequence targeting Luciferase

DOWNLOAD MSDS (PDF)

Synonym: MISSION® TurboGFP® Control Transduction Particles

  •  Council of Europe no. 12000000

  • Popular Documents:  Bulletin (PDF)  

Properties

Related Categories Functional Genomics and RNAi, MISSION Controls in Lentiviral Format, MISSION shRNA, MISSION shRNA Controls
shipped in   dry ice
storage temp.   −70°C

Description

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.

General description

Sigma® is proud to offer IPTG-inducible vectors as the latest development from our continued partnership in TRC.
The pLKO vector has been redesigned to contain a LacI (repressor) and a modified human U6 shRNA promoter with LacO (operator) sequences. In the absence of IPTG (isopropyl-Β-D-thio-galactoside), an analogue of lactose, LacI binds to LacO preventing expression of the shRNA. When IPTG is present, the allosteric LacI repressor changes conformation, releasing itself from lacO modified human U6 promoter, and subsequently allows expression of the shRNA.
The MISSION 1x LacO Inducible Luciferase shRNA Control Transduction Particles contain an shRNA sequence targeting Luciferase, Photinus pyralis (GenBank Accession No. M15077). The Luciferase shRNA Control Particles are useful as a positive knockdown control in experiments using cell lines expressing firefly luciferase.
Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. In addition, the control transduction particles are pseudotyped with an envelope G glycoprotein from Vesicular Stomatitis Virus (VSV-G), allowing transduction of a wide variety of mammalian cells. 200 μl of 106 TU/ml (via p24 titering assay) lentiviral particles are provided as frozen stock.

When conducting experiments using MISSION® shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION Control Transduction Particles are a critical positive control to monitor transduction efficiency.
To see more application data, protocols, vector maps visit sigma.com/shrna.

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?