V5250 Sigma

Snake venom from Bothrops alternata

DOWNLOAD MSDS (PDF)

Synonym: Urutu

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
storage temp.   −20°C

Description

Application

Snake venom from Bothrops alternata (Urutu) may be used as a source of Bothrops venom associated toxins useful in research. It may also be used as an immunogen or for proteome research.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?