Synonym: Anti-WBSCR10, Anti-WBSCR9, Anti-WSTF, Anti-bromodomain adjacent to zinc finger domain, 1B
MDL number MFCD08705534
| Related Categories | 5, AA - CA, Alphabetical Index, Antibodies, Antibodies for Chromatin Remodeling, |
| species reactivity | human |
| application(s) | indirect ELISA: suitable |
| clone | 5E9, monoclonal |
| antibody form | purified immunoglobulin |
| form | buffered aqueous solution |
| isotype | IgG2aκ |
| immunogen sequence |
MDFQTVQNKCSCGSYRSVQEFLTDM |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... BAZ1B(9031) |
BAZ1B (NP_075381, a.a. 1384-1484) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.
Solution in phosphate buffered saline, pH 7.2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?