Synonym: spectroscopy uv visible, uv vis spectroscopy, uvltraviolet spectrophotometer
| Related Categories | Analytical Reagents, Analytical/Chromatography, Labware, Manufacturer Browser, Photometers, |
Features a sleek, new design with up-to-date electronics and the latest optical components. Measures absorbance or transmittance over a range of 325-1100 nm and can be programmed to print out concentrations. Uses 10 mm square cuvettes or 10 mm test tubes. Includes single-position cell holder, 5 disposable cuvettes, RS-232C interface, Centronics-compatible printer port, dust cover, and owner′s manual. 100-240 V compatible.
Customers Also Viewed
for evaluation of the COD cuvet tests see 37735, 37736, 37737, 37738