| Related Categories | Absorption Cells, Analytical Reagents, Analytical/Chromatography, Cuvettes, Hellma, |
| material | Suprasil® quartz |
| description | with 2 polished windows |
| mfr. no. | Hellma 105B-QS |
| limit | 200-2,500 nm spectral range |
| H × W × D | 25 mm × 12.5 mm × 12.5 mm |
| chamber volume | 300 μL |
| pathlength | 10 mm |
with PTFE lid, black side walls and base
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?